Learn More
Meso Scale Discovery UPLEX HUM Galectin9 ASY
SDP
Supplier:  Meso Scale Discovery K151AGWK4
Encompass
The Meso Scale Discovery U-PLEX Human Galectin-9 Assay is an electrochemiluminescent multiplex immunoassay designed to measure Galectin-9 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary U-PLEX technology allowing simultaneous measurement of multiple biomarkers per well in a standard 96-well plate format The U-PLEX Galectin-9 Assay provides high sensitivity and a broad dynamic range while only requiring a small sample volume This assay can aid in understanding immune response regulation and inflammation mediated by Galectin-9
Catalog No. 50-000-81645
Add to Cart
Request a Quote