Learn More
Meso Scale Discovery UPLEX HUM Ghrelin act ASY384
SDP
Supplier:  Meso Scale Discovery K2516KK4
Encompass
The Meso Scale Discovery U-PLEX Human Ghrelin (Active) 384-Well Assay Kit is a multiplex immunoassay that measures active ghrelin levels in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of active ghrelin The assay has a detection range of 0 06-1500 pg/mL and includes a plate coated with capture antibodies detection antibodies calibrators and controls This quantitative immunoassay allows sensitive accurate and rapid analysis of active ghrelin levels from small sample volumes
Catalog No. 50-000-80782
Only null left
Add to Cart
Request a Quote