Learn More
Meso Scale Discovery UPLEX HUM Ghrelin act ASY384
SDP
Supplier:  Meso Scale Discovery K2516KK2
Encompass
The Meso Scale Discovery U-PLEX Human Ghrelin (Active) 384-Well Assay Kit is a multiplex immunoassay that measures active ghrelin levels in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of active ghrelin The assay has a detection range of 0 08-400 pg/mL and includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls This quantitative immunoassay allows sensitive accurate and rapid measurement of active ghrelin levels for research applications
Catalog No. 50-000-80877
Add to Cart
Request a Quote