Learn More
Meso Scale Discovery UPLEX HUM GIP active ASY384
SDP
Supplier:  Meso Scale Discovery K2516NK2
Encompass
The U-PLEX Human GIP Active 384-Well Assay from Meso Scale Discovery is a 384-well electrochemiluminescent immunoassay designed to measure active glucagon-like peptide-1 (GIP) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of active GIP The assay has a lower limit of quantification of 0 061 pg/mL and an upper limit of quantification of 1000 pg/mL It requires just 50 L of sample per well and can be read on MSD s SECTOR Imager 6000 The kit includes a 384-well plate coated with capture antibody detection antibody calibrator and assay diluent
Catalog No. 50-000-78497
Add to Cart
Request a Quote