Learn More
Meso Scale Discovery UPLEX HUM GIP active ASY384
SDP
Supplier:  Meso Scale Discovery K2516NK4
Encompass
The Meso Scale Discovery U-PLEX Human GIP Active 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of active glucagon-like peptide-1 (GIP) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 0 07 pg/mL and an upper limit of quantification of 2500 pg/mL It requires just 25 L of sample per well and includes reagents for a 384-well plate
Catalog No. 50-000-80813
Add to Cart
Request a Quote