Learn More
Meso Scale Discovery UPLEX HUM GIP inactive ASY384
SDP
Supplier:  Meso Scale Discovery K2515TK4
Encompass
The Meso Scale Discovery U-PLEX Human GIP Inactive 384-Well Assay is an electrochemiluminescent immunoassay for measuring human gastric inhibitory polypeptide (GIP) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection with a detection range of 0 08 to 2500 pg/mL The assay processes up to 384 samples with 25 L each in about 3 5 hours offering an efficient workflow for high-throughput GIP quantification
Catalog No. 50-000-80904
Only null left
Add to Cart
Request a Quote