Learn More
Meso Scale Discovery UPLEX HUM GIP inactive ASY384
SDP
Supplier:  Meso Scale Discovery K2515TK2
Encompass
The Meso Scale Discovery U-PLEX Human GIP Inactive 384-Well Assay is a 384-well electrochemiluminescent immunoassay designed to measure inactive GIP in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a 384-well plate coated with a GIP capture antibody detection antibody conjugated to an MSD SULFO-TAG calibrators and buffers for 384 tests The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It requires 2 5 hours total assay time
Catalog No. 50-000-81099
Only null left
Add to Cart
Request a Quote