Learn More
Meso Scale Discovery UPLEX HUM GIP TTL ASY384well
SDP
Supplier:  Meso Scale Discovery K2515SK4
Encompass
The Meso Scale Discovery U-PLEX Human GIP Total 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human gastric inhibitory polypeptide (GIP) in serum plasma and cell culture supernatants It includes a plate with capture antibody detection antibody with an electrochemiluminescent label calibrator and buffer Sensitivity is 0 08 pg/mL with a range of 0 08-1000 pg/mL It uses 25 L samples and takes about 3 5 hours The kit provides reagents for one 384-well plate ensuring accurate and sensitive GIP measurement
Catalog No. 50-000-80785
Only null left
Add to Cart
Request a Quote