Learn More
Meso Scale Discovery UPLEX HUM GITR/TNFRSF18 ASY384
SDP
Supplier:  Meso Scale Discovery K251Y7K2
Encompass
The Meso Scale Discovery U-PLEX Human GITR (TNFRSF18) 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human GITR in cell culture supernatants serum plasma and other biological matrices It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput analysis of GITR levels with minimal hands-on time It includes a GITR-specific capture antibody detection antibody and calibrator
Catalog No. 50-000-79402
Only null left
Add to Cart
Request a Quote