Learn More
Meso Scale Discovery UPLEX HUM GITR/TNFRSF18 ASY384
SDP
Supplier:  Meso Scale Discovery K251Y7K4
Encompass
The GITR/TNFRSF18 ELISA Kit from Meso Scale Discovery is a sandwich immunoassay for the quantitative measurement of GITR in human serum plasma cell culture supernatants cell and tissue lysates The kit uses electrochemiluminescence detection technology and provides high sensitivity wide dynamic range and low sample volume requirements The assay requires 2 5 hours total time and has a lower detection limit of 19 5 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer sample diluent calibrator diluent wash buffer and a calibrator
Catalog No. 50-000-80695
Only null left
Add to Cart
Request a Quote