Learn More
Meso Scale Discovery UPLEX HUM GITRL/TNFSF18 ASY
SDP
Supplier:  Meso Scale Discovery K151S7K4
Encompass
The Meso Scale Discovery U-PLEX Human GITRL (TNFSF18) Assay is a multiplex immunoassay that measures human GITRL in serum plasma and cell culture supernatants using MSD s electrochemiluminescent technology on the MESO QuickPlex SQ 120 It features high sensitivity a wide dynamic range and requires 50 L sample volume The assay detects as low as 19 5 pg/mL and quantifies up to 10 000 pg/mL delivering results in under 3 hours It s suitable for immune monitoring and biomarker research
Catalog No. 50-000-81706
Add to Cart
Request a Quote