Learn More
Meso Scale Discovery UPLEX HUM GITRL/TNFSF18 ASYQP
SDP
Supplier:  Meso Scale Discovery K151S7K24
Encompass
The Meso Scale Discovery U-PLEX Human GITRL (TNFSF18) Assay QuickPlex Kit is a multiplex immunoassay for measuring human GITRL in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection in a quickplex format analyzing 25 samples with 25 L per well The detection limit is 0 04 ng/mL and the quantification limit is 2500 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies and calibrator solutions for a standard curve
Catalog No. 50-000-79831
Add to Cart
Request a Quote