Learn More
Meso Scale Discovery UPLEX HUM GLP1 active ASYQP
SDP
Supplier:  Meso Scale Discovery K1516LK22
Encompass
The MSD U-PLEX Human GLP-1 (Active) Singleplex Assay is an electrochemiluminescence-based immunoassay designed to quantitatively measure active glucagon-like peptide-1 levels in human biological samples This singleplex format allows focused detection of GLP-1 a key incretin hormone involved in glucose homeostasis and diabetes research The assay utilizes MSD s proprietary U-PLEX platform technology for sensitive and specific biomarker analysis in clinical and research applications
Catalog No. 50-000-80291