Learn More
Meso Scale Discovery UPLEX HUM GLPactive ASY384
SDP
Supplier:  Meso Scale Discovery K2516LK4
Encompass
The Meso Scale Discovery U-PLEX Human GLP-1 (Active) 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of active glucagon-like peptide-1 (GLP-1) in human serum plasma and cell culture supernatants The assay utilizes MSD GOLD technology and provides high sensitivity broad dynamic range and minimal matrix interference The kit includes a 384-well plate coated with capture antibody detection antibody read buffer calibrator and controls for the quantitative analysis of active GLP-1 between 0 15-1000 pg/mL when using a 10 L sample size The assay can be used to study GLP-1 biology and for biomarker research
Catalog No. 50-000-81104
Add to Cart
Request a Quote