Learn More
Meso Scale Discovery UPLEX HUM GLPactive ASY384well
SDP
Supplier:  Meso Scale Discovery K2516LK2
Encompass
The Meso Scale Discovery U-PLEX Human GLP-1 (Active) 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring active GLP-1 in human serum plasma and cell culture supernatants It uses MSD GOLD technology for high sensitivity and low sample volume The kit includes a plate with capture antibodies detection antibodies and calibrators for a standard curve quantifying GLP-1 between 0 061-1500 pg/mL It processes 14 samples in triplicate with results read on the MESO QuickPlex SQ 120 This is a sensitive high-throughput kit for quantifying active GLP-1
Catalog No. 50-000-80984
Only null left
Add to Cart
Request a Quote