Learn More
Meso Scale Discovery UPLEX HUM Glucagon ASY384well
SDP
Supplier:  Meso Scale Discovery K2515YK2
Encompass
The Meso Scale Discovery U-PLEX Human Glucagon 384-Well Assay Kit is a multiplex immunoassay that measures glucagon levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate format for high throughput analysis The assay has a detection range of 1 56 - 400 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator solutions It can be used to study glucagon biology and for drug discovery research related to diabetes obesity and metabolic disorders The kit includes sufficient reagents for one 384-well plate
Catalog No. 50-000-80908
Add to Cart
Request a Quote