Learn More
Meso Scale Discovery UPLEX HUM gp130 soluble ASY384
SDP
Supplier:  Meso Scale Discovery K251P7K2
Encompass
The Meso Scale Discovery U-PLEX Human GP130 Soluble 384-Well Assay is a 384-well multiplex immunoassay kit that measures soluble GP130 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay allows for the simultaneous measurement of GP130 along with other biomarkers in the same sample It requires an MSD instrument such as SECTOR Imager 2400 or 6000 for analysis and data acquisition With a measuring range of 0 05-1500 pg/mL this assay kit enables the reliable quantification of GP130
Catalog No. 50-000-79568
Only null left
Add to Cart
Request a Quote