Learn More
Meso Scale Discovery UPLEX HUM Granzyme A ASYQP
SDP
Supplier:  Meso Scale Discovery K151G8K24
Encompass
The Meso Scale Discovery U-PLEX Human Granzyme A Assay Kit is designed for the quantitative measurement of Granzyme A in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD plate technology coated with capture antibodies and includes a vial of SULFO-TAG labeled detection antibodies The kit includes standards ranging from 3 2 pg/mL to 10 000 pg/mL and can detect Granzyme A with a lower limit of detection of 0 3 pg/mL This assay allows high sensitivity quantitative detection of Granzyme A from small sample volumes
Catalog No. 50-000-78776
Only null left
Add to Cart
Request a Quote