Learn More
Meso Scale Discovery UPLEX HUM HAVCR2/TIM3 ASYQP
SDP
Supplier:  Meso Scale Discovery K151E8K24
Encompass
The Meso Scale Discovery U-PLEX Human HAVCR2 (TIM-3) Assay is a multiplex immunoassay kit for detecting and quantifying human TIM-3 and other biomarkers in serum plasma and cell culture supernatants It uses MSD s MULTI-ARRAY technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators Results are analyzed with MSD s QuickPlex SQ 120 This assay efficiently measures TIM-3 important in immune response regulation
Catalog No. 50-000-80194
Only null left
Add to Cart
Request a Quote