Learn More
Meso Scale Discovery UPLEX HUM HVEM/TNFRSF14 ASY384
SDP
Supplier:  Meso Scale Discovery K251AGXK4
Encompass
The Meso Scale Discovery Human HVEM/TNFRSF14 Kit is an electrochemiluminescent immunoassay designed for the quantitative measurement of human herpesvirus entry mediator (HVEM) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels which emit light when electrochemically stimulated via carbon electrodes in microplates The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrator and sample diluent The assay has a typical sensitivity of 19 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80988
Only null left
Add to Cart
Request a Quote