Learn More
Meso Scale Discovery UPLEX HUM HVEM/TNFRSF14 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AGXK21
Encompass
The Meso Scale Discovery U-PLEX Human HVEM (TNFRSF14) Assay is a multiplex immunoassay kit for detecting and quantifying human HVEM in various biological matrices It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results with a detection range of 0 05-1000 pg/mL The assay requires an MSD instrument like QuickPlex to read the plate With high sensitivity broad dynamic range and minimal hands-on time this kit allows efficient and accurate HVEM analysis
Catalog No. 50-000-81646