Learn More
Meso Scale Discovery UPLEX HUM I309 ASY384well
SDP
Supplier:  Meso Scale Discovery K251UYK4
Encompass
The Meso Scale Discovery U-PLEX Human I-309/384-Well Assay is a 384-well multiplex immunoassay kit that enables simultaneous quantification of human I-309 and 384 other biomarkers It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay requires only 25 L of sample per well and can perform up to 40 plates per kit It is suitable for serum plasma and cell culture supernatants The kit includes a 384-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and wash buffer
Catalog No. 50-000-80702
Only null left
Add to Cart
Request a Quote