Learn More
Meso Scale Discovery UPLEX HUM ICAM1 ASY384well
SDP
Supplier:  Meso Scale Discovery K251E9K4
Encompass
The Meso Scale Discovery U-PLEX Human ICAM-1 384-Well Assay Kit is a multiplex immunoassay that measures Intercellular Adhesion Molecule 1 (ICAM-1) in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption It is available in a 384-well plate format includes all necessary reagents and can perform 39 tests in duplicate This assay has been validated for measuring ICAM-1 levels in disease research biomarker discovery and drug development
Catalog No. 50-000-80752
Add to Cart
Request a Quote