Learn More
Meso Scale Discovery UPLEX HUM ICOS ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151AHBK24
Encompass
The Meso Scale Discovery U-PLEX Human ICOS Assay is a multiplex immunoassay that measures human ICOS in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows simultaneous measurement of ICOS and other biomarkers It consists of a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and an MSD reader The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-80472
Only null left
Add to Cart
Request a Quote