Learn More
Meso Scale Discovery UPLEX HUM ICOSL/B7H2 ASY
SDP
Supplier:  Meso Scale Discovery K151AHJK4
Encompass
The Meso Scale Discovery U-PLEX Human ICOS/L B7-H2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Inducible T cell COStimulator (ICOS) and B7 Homolog 2 (B7-H2) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for efficient and accurate analysis of ICOS and B7-H2 which play important roles in immune regulation
Catalog No. 50-000-80474
Only null left
Add to Cart
Request a Quote