Learn More
Meso Scale Discovery UPLEX HUM ICOSL/B7H2 ASY384
SDP
Supplier:  Meso Scale Discovery K251AHJK2
Encompass
The Meso Scale Discovery U-PLEX Human ICOS/L B7-H2 Assay 384-well kit is a multiplex immunoassay that measures human ICOS and B7-H2 in cell culture supernatants serum plasma and other biological samples It uses electrochemiluminescence detection technology in a 384-well plate format allowing high-throughput analysis of up to 80 samples in duplicate The assay takes approximately 3 hours to complete and has a detection range of 0 05-2500 pg/mL for ICOS and 0 03-1500 pg/mL for B7-H2 This quantitative immunoassay kit provides sensitive accurate and reproducible results for measuring levels of human ICOS and B7-H2
Catalog No. 50-000-78662
Add to Cart
Request a Quote