Learn More
Meso Scale Discovery UPLEX HUM ICOSL/B7H2 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHJK24
Encompass
The Meso Scale Discovery U-PLEX Human ICOS/L B7-H2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Inducible T cell COStimulator (ICOS) and B7 Homolog 2 (B7-H2) biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve This assay provides reproducible and sensitive quantification of ICOS and B7-H2 useful biomarkers for monitoring immune system function
Catalog No. 50-000-78803
Add to Cart
Request a Quote