Learn More
Meso Scale Discovery UPLEX HUM IFN 2a ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151VHK4
Encompass
The Meso Scale Discovery U-PLEX Human IFN-alpha2a Assay Kit is designed to quantitatively measure human interferon alpha 2a (IFN-alpha2a) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection method The assay uses MSD GOLD Streptavidin Multi-Array 96-well plates and U-PLEX linker to capture and detect IFN-alpha2a using a sandwich immunoassay format The kit includes a vial of lyophilized IFN-alpha2a calibrator to generate a standard curve and vials of detection antibody and read buffer The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 400 pg/mL for IFN-alpha2a
Catalog No. 50-000-79543
Only null left
Add to Cart
Request a Quote