Learn More
Meso Scale Discovery UPLEX HUM IFN 2a ASY384well
SDP
Supplier:  Meso Scale Discovery K251VHK2
Encompass
The Meso Scale Discovery U-PLEX Human IFN-alpha2a 384-Well Assay Kit is designed to measure human interferon alpha 2a (IFN-alpha2a) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection method The kit includes a 384-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 400 pg/mL The kit can perform up to 380 samples in 384-well format with a sample volume of 25 L
Catalog No. 50-000-79986
Add to Cart
Request a Quote