Learn More
Meso Scale Discovery UPLEX HUM IFN ASY384well
SDP
Supplier:  Meso Scale Discovery K251TTK4
Encompass
The Meso Scale Discovery U-PLEX Human IFN gamma 384-Well Assay is an electrochemiluminescent immunoassay for quantifying IFN gamma in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for a sensitive reproducible assay in a 384-well format Sensitivity is 0 04 pg/mL with an upper limit of 2500 pg/mL It requires 25 L of sample and takes about 3 5 hours The kit includes a 384-well plate detection antibodies buffers calibrators and controls suitable for high-throughput use
Catalog No. 50-000-80727
Only null left
Add to Cart
Request a Quote