Learn More
Meso Scale Discovery UPLEX HUM IFN ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151TTK24
Encompass
The U-PLEX Human IFN Assay (K151TTK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of interferon-gamma in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX technology enables researchers to measure cytokine levels in serum plasma and cell culture supernatants with superior precision and reproducibility compared to traditional ELISA methods
Catalog No. 50-000-78522
Only null left
Add to Cart
Request a Quote