Learn More
Meso Scale Discovery UPLEX HUM IL1 ASY384well
SDP
Supplier:  Meso Scale Discovery K251UNK2
Encompass
The Meso Scale Discovery U-PLEX Human IL-1alpha 384-well Assay Kit is a multiplex immunoassay that measures human interleukin 1 alpha (IL-1alpha) in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay is provided in a 384-well plate format and includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The kit allows quantification of IL-1alpha concentrations and can be used for biomarker discovery and validation
Catalog No. 50-000-80722
Add to Cart
Request a Quote