Learn More
Meso Scale Discovery UPLEX HUM IL1 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151UNK24
Encompass
The U-PLEX Human IL-1 Assay (K151UNK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-1 alpha levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for cytokine detection supporting research applications in inflammation immunology and disease biomarker studies The kit provides reliable reproducible results for laboratory research use
Catalog No. 50-000-78244
Add to Cart
Request a Quote