Learn More
Meso Scale Discovery UPLEX HUM IL12/IL23p40 ASY
SDP
Supplier:  Meso Scale Discovery K151UQK1
Encompass
The Meso Scale Discovery U-PLEX Human IL-12/IL-23p40 Assay Kit is a multiplex immunoassay that measures IL-12 and IL-23 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The kit has a typical sensitivity of 0 3 pg/mL and an upper limit of 50 000 pg/mL
Catalog No. 50-000-80396