Learn More
Meso Scale Discovery UPLEX HUM IL12/IL23p40 ASYQP
SDP
Supplier:  Meso Scale Discovery K151UQK21
Encompass
The U-PLEX Human IL-12/IL-23p40 Assay (K151UQK) is a multiplex immunoassay from Meso Scale Discovery that simultaneously measures interleukin-12 and the p40 subunit shared by IL-12 and IL-23 in human samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing The assay is designed for research applications studying inflammatory responses and immune system function offering reliable quantification of these critical cytokines in serum plasma and cell culture supernatants
Catalog No. 50-000-79147