Learn More
Meso Scale Discovery UPLEX HUM IL12p70 ASY384well
SDP
Supplier:  Meso Scale Discovery K251UAK2
Encompass
The Meso Scale Discovery U-PLEX Human IL-12p70 384-Well Assay Kit is a multiplex immunoassay that measures IL-12p70 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It allows simultaneous measurement of IL-12p70 levels from a small sample volume in under 3 hours The kit has high specificity and low cross-reactivity
Catalog No. 50-000-80689
Only null left
Add to Cart
Request a Quote