Learn More
Meso Scale Discovery UPLEX HUM IL15 ASY384well
SDP
Supplier:  Meso Scale Discovery K251URK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-15 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 15 (IL-15) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents It has a detection range of 1 53-2500 pg/mL and no cross-reactivity with other cytokines The assay takes about 3 5 hours
Catalog No. 50-000-80972
Add to Cart
Request a Quote