Learn More
Meso Scale Discovery UPLEX HUM IL17A ASY384well
SDP
Supplier:  Meso Scale Discovery K251UTK2
Encompass
The Meso Scale Discovery U-PLEX Human IL-17A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-17A in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager 6000 for high sensitivity detection The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and diluent The assay has a detection range of 0 06-1500 pg/mL and takes approximately 3 5 hours to complete This highly sensitive and reproducible assay allows rapid quantification of IL-17A
Catalog No. 50-000-80511
Only null left
Add to Cart
Request a Quote