Learn More
Meso Scale Discovery UPLEX HUM IL17A/F ASY384well
SDP
Supplier:  Meso Scale Discovery K251VYK2
Encompass
The Meso Scale Discovery U-PLEX Human IL-17A/F 384-Well Assay is a multiplex immunoassay that measures human IL-17A and IL-17F in serum plasma cell culture supernatants and other biological samples It utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay has a typical sensitivity of 0 04-0 74 pg/mL and requires 12 5 L of sample per well It includes a 384-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance Turnaround time is 3 5 hours
Catalog No. 50-000-80619
Add to Cart
Request a Quote