Learn More
Meso Scale Discovery UPLEX HUM IL17C ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151WJK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-17C Assay Kit is a multiplex immunoassay that measures IL-17C levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81478
Add to Cart
Request a Quote