Learn More
Meso Scale Discovery UPLEX HUM IL17D ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151XOK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-17D Assay Kit is a multiplex immunoassay that measures human IL-17D levels in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve and validate assay performance The assay takes approximately 3 5 hours to complete and is designed for use on MSD instrumentation including the MESO QuickPlex SQ 120
Catalog No. 50-000-79708
Add to Cart
Request a Quote