Learn More
Meso Scale Discovery UPLEX HUM IL17E/IL25 ASY384
SDP
Supplier:  Meso Scale Discovery K251VZK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human interleukin 17E (IL-17E) and interleukin 25 (IL-25) biomarkers using electrochemiluminescence detection technology The assay utilizes Meso Scale Discovery s proprietary multi-array plate technology and provides reagents for 384 tests in a ready-to-use kit format The assay has a testing time of 3 5 hours and requires 2 5 hours of hands-on time The lower limits of detection are 0 04 pg/mL for IL-17E and 0 17 pg/mL for IL-25 The assay is designed for research use only and is not intended for diagnostic procedures
Catalog No. 50-000-80175
Add to Cart
Request a Quote