Learn More
Meso Scale Discovery UPLEX HUM IL17F ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151WAK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-17F Assay Kit is a multiplex immunoassay that measures IL-17F levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81473
Only null left
Add to Cart
Request a Quote