Learn More
Meso Scale Discovery UPLEX HUM IL1B ASY384well
SDP
Supplier:  Meso Scale Discovery K251TUK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-1beta 384-Well Assay Kit is a multiplex immunoassay that measures IL-1beta levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent It allows simultaneous measurement of IL-1beta along with other biomarkers in the U-PLEX platform The kit has high performance and is designed for biomarker research and drug discovery applications
Catalog No. 50-000-80686
Only null left
Add to Cart
Request a Quote