Learn More
Meso Scale Discovery UPLEX HUM IL1B ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151TUK24
Encompass
The MSD U-PLEX Human IL-1 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative measurement of interleukin-1 beta in human biological samples This singleplex format allows focused analysis of IL-1 a key pro-inflammatory cytokine involved in immune responses and inflammatory processes The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology inflammation studies and biomarker discovery
Catalog No. 50-000-81987
Only null left
Add to Cart
Request a Quote