Learn More
Meso Scale Discovery UPLEX HUM IL33 ASY384well
SDP
Supplier:  Meso Scale Discovery K251WFK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-33 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of IL-33 in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a sensitivity of 0 03 pg/mL and a broad dynamic range from 0 03-1000 pg/mL It requires a low sample volume of 25 L and the total assay time is under 3 hours The kit includes reagents for 384 tests and can be used to measure IL-33 on the MESO QuickPlex SQ 120
Catalog No. 50-000-81128
Add to Cart
Request a Quote