Learn More
Meso Scale Discovery UPLEX HUM IL6 ASY384well
SDP
Supplier:  Meso Scale Discovery K251TXK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-6 Assay Kit is a multiplex immunoassay that measures IL-6 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 1500 pg/mL
Catalog No. 50-000-81043
Add to Cart
Request a Quote