Learn More
Meso Scale Discovery UPLEX HUM IL9 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151XKK24
Encompass
The U-PLEX Human IL-9 Assay (K151XKK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of interleukin-9 protein levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible customizable multiplex panels that can be combined with other biomarkers for comprehensive cytokine profiling in research applications
Catalog No. 50-000-81815
Only null left
Add to Cart
Request a Quote