Learn More
Meso Scale Discovery UPLEX HUM Insulin ASY384well
SDP
Supplier:  Meso Scale Discovery K2516HK2
Encompass
The Meso Scale Discovery U-PLEX Human Insulin 384-Well Assay Kit is a multiplex immunoassay that measures insulin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label calibrators and controls It allows simultaneous measurement of insulin levels from 16 samples per plate The assay has a typical lower limit of detection of 0 058 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80983
Add to Cart
Request a Quote