Learn More
Meso Scale Discovery UPLEX HUM LIGHT/TNFSF14 ASY
SDP
Supplier:  Meso Scale Discovery K151AHCK4
Encompass
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 (TNFSF14) Assay is a multiplex immunoassay that measures human TNFSF14 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows for the simultaneous measurement of TNFSF14 along with other biomarkers It requires 50 L of sample and can be read on MSD instrument platforms The lower limit of detection for TNFSF14 is 0 04 pg/mL
Catalog No. 50-000-78405
Add to Cart
Request a Quote